Learn More
Abcam Dylight 350 Conjugation Kit (Fast) - Lightning-Link, 1MG
Catalog No. 502589152
Encompass_Preferred
Labeling in < 20 mins with 30 secs hands-on time using Dylight 350 Conjugation Kit (Fast) - Lightning-Link ab201797. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9152 Supplier Abcam Supplier No. AB2017971MG
Labeling in < 20 mins with 30 secs hands-on time using Dylight 350 Conjugation Kit (Fast) - Lightning-Link ab201797. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement