Learn More
Jackson Immuno Research Labs DYLIGHT 405 ANTI MOUSE IGG 1MG
SDP
Supplier:  Jackson Immuno Research Labs 115475146
Encompass_Preferred
DyLight 405-AffiniPure Goat Anti-Mouse IgG (H+L) (min X Hu,Bov,Hrs,Rb,Sw Sr Prot)
Catalog No. NC0328758