Learn More
Vector Laboratories Goat Anti-Rabbit IgG Antibody DyLight 488 1.5 mg
SDP
Supplier:  Vector Laboratories DI14881.5
Encompass
Vector Laboratories DyLight®488 Goat Anti-Rabbit IgG Antibody is thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. There is minimal cross-reactivity to human, mouse, horse, cow, pig, cat, dog, chicken, monkey, guinea pig, hamster, rat and sheep. The excitation and emission spectra of DyLight® fluorescent dyes parallel those of other commercially available fluorescent reagents, allowing for easy substitution into existing protocols. The fluorescent dye is conjugated in a way that ensures the maximum degree of labeling without compromising the specificity or affinity of the antibody. DyLight 488 Anti-Rabbit IgG Antibody is supplied in liquid format and recognizes both heavy and light chains. The recommended concentration range for most applications is 5-20 micrograms/ml. Excitation: 493 nm, Emission: 518 nm, Color: Green.
Catalog No. NC0696900