Learn More
Abcam DyLight 550 Conjugation Kit (Fast) - Lightning-Link, 3 x 100UG
Catalog No. 502589141
Encompass_Preferred
DyLight 550 conjugation / labeling in < 20 mins with 30 secs hands-on time using DyLight 550 Conjugation Kit (Fast) - Lightning-Link ab201800. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9141 Supplier Abcam Supplier No. AB2018003X100UG
DyLight 550 conjugation / labeling in < 20 mins with 30 secs hands-on time using DyLight 550 Conjugation Kit (Fast) - Lightning-Link ab201800. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement