Learn More
Cell Signaling Technology DyLight™ 554 Phalloidin, Cytoskeleton Labeling, 300 Units
SDP
Catalog No. NC0595737
Encompass
DyLight™ 554 Phalloidin is a fluorescent probe designed for labeling F-actin in fixed cells. It utilizes a high-affinity interaction with F-actin to provide clear visualization of the cytoskeleton in various immunofluorescence applications. This product is supplied as a lyophilized solid and requires reconstitution in anhydrous DMSO before use.
  • Excitation and emission wavelengths of 551 nm and 572 nm, respectively
  • Stable for 1 year when stored at -20°C, desiccated and protected from light
  • Suitable for immunofluorescence in fixed cells and frozen tissue sections
  • Provides enough material for 3,000 assays when used at a 1:400 dilution in a 100 μl volume
  • Recommended for use with anhydrous DMSO reconstitution only
Catalog No. NC0595737 Supplier Cell Signaling Technology Supplier No. 13054S
DyLight™ 554 Phalloidin is a fluorescent probe designed for labeling F-actin in fixed cells. It utilizes a high-affinity interaction with F-actin to provide clear visualization of the cytoskeleton in various immunofluorescence applications. This product is supplied as a lyophilized solid and requires reconstitution in anhydrous DMSO before use.
  • Excitation and emission wavelengths of 551 nm and 572 nm, respectively
  • Stable for 1 year when stored at -20°C, desiccated and protected from light
  • Suitable for immunofluorescence in fixed cells and frozen tissue sections
  • Provides enough material for 3,000 assays when used at a 1:400 dilution in a 100 μl volume
  • Recommended for use with anhydrous DMSO reconstitution only