Learn More
Vector Laboratories Griffonia Simplicifolia Lectin -Isolectin B4 DyLight 594 0.5 mg
SDP
Catalog No. NC0173303
Encompass
GSL I-B4 isolectin is a useful marker for endothelial cells from nonprimates such as mouse, rat, rabbit and goat, as well as a marker for non-peptidergic unmyelinated primary afferent neurons. This "B"-rich lectin preferentially agglutinates blood group B cells and is specific for α-galactose residues. Produced using special conjugation procedures to incorporate DyLight 594 into our affinity-purified lectin, the ratio of fluorophore to lectin protein provides optimal staining (excitation 592 nm, emission 617 nm). Supplied as a solution and essentially free of unconjugated fluorophores, it is preserved with sodium azide.
Catalog No. NC0173303 Supplier Vector Laboratories Supplier No. DL1207.5
GSL I-B4 isolectin is a useful marker for endothelial cells from nonprimates such as mouse, rat, rabbit and goat, as well as a marker for non-peptidergic unmyelinated primary afferent neurons. This "B"-rich lectin preferentially agglutinates blood group B cells and is specific for α-galactose residues. Produced using special conjugation procedures to incorporate DyLight 594 into our affinity-purified lectin, the ratio of fluorophore to lectin protein provides optimal staining (excitation 592 nm, emission 617 nm). Supplied as a solution and essentially free of unconjugated fluorophores, it is preserved with sodium azide.