Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyLight™ 594 NHS Ester

Catalog No. PI46413 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
5 x 65 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI46413 5 x 65 μg
PI46412 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI46413 Supplier Thermo Scientific™ Supplier No. 46413
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of NHS ester-activated DyLight Dyes.

Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.

The DyLight Dyes exhibit higher fluorescence intensity and photostability than Alexa™, CyDye™ and LI-COR™ Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9). Additionally, the water solubility of the DyLight Dyes allows a high dye-to-protein ratio without precipitation during conjugation.

Highlights:

  • Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate

  • Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)

  • Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 65 μg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C.
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye DyLight™ 594
Label Type DyLight Fluors
Product Line DyLight
Emission 618
Excitation 593
Labeling Target Proteins
Show More Show Less
Can you provide the shelf-life for DyLight 594 NHS Ester?

DyLight 594 NHS Ester is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.