Learn More
Abcam DyLight 755 Conjugation Kit (Fast) - Lightning-Link, 1MG
Catalog No. 502589124
Encompass_Preferred
Labeling in < 20 mins with 30 secs hands-on time using DyLight 755 Conjugation Kit (Fast) - Lightning-Link ab201805. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9124 Supplier Abcam Supplier No. AB2018051MG
Labeling in < 20 mins with 30 secs hands-on time using DyLight 755 Conjugation Kit (Fast) - Lightning-Link ab201805. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement