Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyLight™ 405 Maleimide

Catalog No. PI46600 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI46600 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI46600 Supplier Thermo Scientific™ Supplier No. 46600
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of maleimide-activated DyLight Dyes.

Thermo Scientific™ Sulfhydryl-Reactive DyLight Dyes are maleimide-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as probes for cellular imaging and other fluorescence detection methods.

The Thermo Scientific™ DyLight™ Dyes have absorption spectra ranging from 350 to 770nm. These reagents fluoresce over a broad pH range, are more intense than Alexa Fluor™ or Cy™ Dyes in many applications and match the output wavelengths of common fluorescence instrumentation. Additionally, the water solubility of the DyLight Reagents allows a high fluor-to-protein ratio without precipitation during conjugation.
The sulfhydryl-reactive dyes contain maleimide groups that react predominantly with free –SH groups at pH 6.5-7.5, forming a stable thioether bond. At pH 7, the maleimide group is ∼1000 times more reactive toward a free sulfhydryl than to an amine.1At pH values > 7.5, reactivity toward primary amines increases and hydrolysis of the maleimide group can occur.

Highlights:
  • Specific — maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
  • Efficient labeling methods — well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
  • Optimized antibody labeling procedure —complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
  • Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
  • Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies
TRUSTED_SUSTAINABILITY

Specifications

Description DyLight 405 Maleimide
Quantity 1 mg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C. Shipped at ambient temperature.
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Label or Dye DyLight™ 405
Label Type DyLight Fluors
Product Line DyLight
Emission 420
Excitation 400
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.