Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology DyLight™ 488 Phalloidin, Lyophilized Solid, 300 Units
SDP

Catalog No. 502050589
Encompass

DyLight™ 488 Phalloidin is a fluorescent probe designed for labeling the cytoskeleton of fixed cells by binding with high affinity to F-actin. Due to the inherent toxicity of phalloidin, this product is intended for use in fixed cells only and is not suitable for live cell imaging. The lyophilized material, when reconstituted, provides sufficient stock solution for 3,000 assays.

  • Excitation: 495 nm; Emission: 521 nm.
  • Stable for 1 year at -20°C when stored desiccated and protected from light.
  • Reconstitution requires anhydrous DMSO; aqueous solution stability is low.
  • Compatible with immunofluorescence in all species.
  • Recommended for use with methanol-free 4% formaldehyde fixation.

Catalog No. 50-205-0589 Supplier Cell Signaling Technology Supplier No. 12935S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DyLight™ 488 Phalloidin is a fluorescent probe designed for labeling the cytoskeleton of fixed cells by binding with high affinity to F-actin. Due to the inherent toxicity of phalloidin, this product is intended for use in fixed cells only and is not suitable for live cell imaging. The lyophilized material, when reconstituted, provides sufficient stock solution for 3,000 assays.

  • Excitation: 495 nm; Emission: 521 nm.
  • Stable for 1 year at -20°C when stored desiccated and protected from light.
  • Reconstitution requires anhydrous DMSO; aqueous solution stability is low.
  • Compatible with immunofluorescence in all species.
  • Recommended for use with methanol-free 4% formaldehyde fixation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.