Learn More
Vector Laboratories Goat Anti-Rabbit IgG Antibody DyLight 594 1.5 mg
SDP
Supplier:  Vector Laboratories DI15941.5
Encompass
Vector Laboratories DyLight®594 Goat Anti-Rabbit IgG Antibody is thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. There is minimal cross-reactivity to human, mouse, horse, cow, pig, cat, dog, chicken, monkey, guinea pig, hamster, rat and sheep. The excitation and emission spectra of DyLight fluorescent dyes parallel those of other commercially available fluorescent reagents, allowing for easy substitution into existing protocols. The fluorescent dye is conjugated in a way that ensures the maximum degree of labeling without compromising the specificity or affinity of the antibody. DyLight 594 Anti-Rabbit IgG Antibody is supplied in liquid format and recognizes both heavy and light chains. The recommended concentration range for most applications is 5-20 micrograms/ml. Excitation: 592 nm, Emission: 617 nm, Color: Red.
Catalog No. NC0696899