Learn More
Uline Dymo 450 Twin Turbo Printer Ea
Catalog No. NC2392360
Encompass
Portable thermal label printer prints address, shipping, file folder, name badge & barcode labels at speeds up to 71 labels per minute; connects via USB or serial cable & compatible with DYMO Label v.8 software; includes LCD display screen & function keys for standalone label printing.
Catalog No. NC2392360 Supplier Uline Supplier No. H1266
Portable thermal label printer prints address, shipping, file folder, name badge & barcode labels at speeds up to 71 labels per minute; connects via USB or serial cable & compatible with DYMO Label v.8 software; includes LCD display screen & function keys for standalone label printing.