Learn More
GA International DYMO-Compatible Cryogenic Labels 1.04" x 0.5" + 0.375" (circle) / 26.4mm x 12.7mm + 9.5mm (circle) Rectangle + Circle Violet From -196°C to +70°C (-320.8°F to +158°F)
Catalog No. 5000571165
Encompass_Preferred
DYMO compatible direct thermal cryo labels. For ultra-low and cryogenic temperature applications (as low as -196°C). Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (-80°C, -50°C, -20°C).
Catalog No. 50-005-71165 Supplier GA International Supplier No. EG1F081VI
DYMO compatible direct thermal cryo labels. For ultra-low and cryogenic temperature applications (as low as -196°C). Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (-80°C, -50°C, -20°C).