Learn More
GA International DYMO-Compatible Cryogenic Labels with SimPEEL™ Technology (Patent Pending) 1" x 1" / 25.4mm x 25.4mm Rectangle Grey From -196°C to +70°C (-320.8°F to +158°F)
Catalog No. 5000571283
Encompass_Preferred
DYMO-compatible cryogenic labels, featuring our new SimPEEL™ technology that makes them even easier to peel from their liner. Perfect for identifying small diameter cryo vials, tubes, and boxes, their innovative adhesive-free tab ensures they won’t stick to your glove or hands for hassle-free application. These direct thermal printable labels are designed to withstand long-term storage in low-temperature freezers (-80°C/-112°C), liquid nitrogen tanks (-196°C/-321°F), and for transportation on dry ice. Perfect for tracking samples with 1D and 2D barcodes or identifying vials with serialized data and variable text.*SimPEEL™ can be integrated into most DYMO-Compatible labels. Please contact us for more information.
Catalog No. 50-005-71283 Supplier GA International Supplier No. SPEF1F070GR
DYMO-compatible cryogenic labels, featuring our new SimPEEL™ technology that makes them even easier to peel from their liner. Perfect for identifying small diameter cryo vials, tubes, and boxes, their innovative adhesive-free tab ensures they won’t stick to your glove or hands for hassle-free application. These direct thermal printable labels are designed to withstand long-term storage in low-temperature freezers (-80°C/-112°C), liquid nitrogen tanks (-196°C/-321°F), and for transportation on dry ice. Perfect for tracking samples with 1D and 2D barcodes or identifying vials with serialized data and variable text.*SimPEEL™ can be integrated into most DYMO-Compatible labels. Please contact us for more information.