Learn More
GA International DYMO-Compatible Paper Labels 0.5" / 12.7mm Circle Red Ambient temperature
Catalog No. 5000571521
Encompass_Preferred
DYMO compatible paper labels for general use applications and laboratory organization. Ideal for office use on file folders, boxes, and shipping containers. Not suitable for deep-freeze applications.
Catalog No. 50-005-71521 Supplier GA International Supplier No. ECY050RE
DYMO compatible paper labels for general use applications and laboratory organization. Ideal for office use on file folders, boxes, and shipping containers. Not suitable for deep-freeze applications.