Learn More
GA International DYMO-Compatible Cryogenic Labels 2.125" x 3.976" / 54mm x 101mm Rectangle Orange From -196°C to +70°C (-320.8°F to +158°F)
Supplier:  GA International EF1F083OR
Encompass_Preferred
Our DYMO-compatible cryo labels are available in a range of sizes and configurations, perfect for identifying 0.5 – 5 ml cryo vials and microtubes, vial caps/tops, cryo/freezer boxes, as well as microplates, Petri dishes, and cell culture plates. These cryogenic labels are designed for short and long-term cryopreservation in ultra-low temperature freezers (-80°C/-112°F, -50°C/-58°F, -20°C/-4°F), liquid phase and vapor phase liquid nitrogen (-196°C/-321°F), and transportation on dry ice. These labels will also withstand temperatures up to +70°C (+158°F), ideal for various applications throughout the lab, including use in incubators.Printable with DYMO 450 LabelWriter printers, these direct thermal labels do not require any additional ink ribbons or other ink sources for printing. Most are also available in various colors for color coding, and easy identification. Cryo-DTermo dot labels (0.25” – 0.375”) are compatible with our label applicator tool, PikaTAG.Not suitable for use with
Catalog No. NC1762494