Learn More
MSC Dymo Digital Postal Scale, 25 Lb, 0.01 Lb, 1 ea
Catalog No. 5000782297
Encompass
The digital postal scale weighs envelopes and packages up to 25 lbs (11 kg). It features an LCD screen for easy-to-read digital weight display and offers convenient hold and tare functions. The compact design allows for desk use and easy storage.
  • LCD screen displays weight in pounds/ounces or kilograms/grams
  • Compact design fits on any desk and stores easily
  • Hold feature locks weight display for 10 seconds after item removal
  • Tare function allows weighing items in containers without container weight
  • Connects via USB cable for use with online mailing and shipping services
  • Automatically shuts off when not in use to conserve battery power
  • Operates with dual power options: USB or three AAA alkaline batteries
  • Factory-calibrated and does not require re-calibration
Catalog No. 50-007-82297 Supplier MSC Supplier No. 66335993
The digital postal scale weighs envelopes and packages up to 25 lbs (11 kg). It features an LCD screen for easy-to-read digital weight display and offers convenient hold and tare functions. The compact design allows for desk use and easy storage.
  • LCD screen displays weight in pounds/ounces or kilograms/grams
  • Compact design fits on any desk and stores easily
  • Hold feature locks weight display for 10 seconds after item removal
  • Tare function allows weighing items in containers without container weight
  • Connects via USB cable for use with online mailing and shipping services
  • Automatically shuts off when not in use to conserve battery power
  • Operates with dual power options: USB or three AAA alkaline batteries
  • Factory-calibrated and does not require re-calibration