Learn More
MSC Dymo Digital Scale, 100 Lb, 0.2 Lbs Graduation, 1 ea
Catalog No. 5000803716
Encompass
This digital scale is designed for accurate weight measurement up to 100 Lb, featuring a clear digital display and an automatic shut-off function to conserve battery life.
  • Measures up to 100 Lb
  • Provides 0.2 Lbs graduation
  • Features a digital display
  • Equipped with a 15 in base length
  • Automatic shut-off functionality prolongs battery life
  • Hold feature locks weight display for 10 seconds
  • Includes USB cable
Catalog No. 50-008-03716 Supplier MSC Supplier No. 66655846
This digital scale is designed for accurate weight measurement up to 100 Lb, featuring a clear digital display and an automatic shut-off function to conserve battery life.
  • Measures up to 100 Lb
  • Provides 0.2 Lbs graduation
  • Features a digital display
  • Equipped with a 15 in base length
  • Automatic shut-off functionality prolongs battery life
  • Hold feature locks weight display for 10 seconds
  • Includes USB cable