Learn More
Enterprise Technology Solutions Dymo DY XTL Laminated Wrap, 13/16"x1-9/16", Black/White
SDP
Catalog No. 5000739563
Encompass_Preferred
Glossy wire/cable wraps feature a laminated overwrap to protect your printed areas from oil, chemicals, moisture, abrasion and fading. Thermal transfer printing technology protects from smearing and fading for long-lasting print visibility. Pre-sized design helps facilitate batch label printing to improve productivity. Wire/cable wraps are perfect for flat surface labeling, such as bar codes, faceplates, shelves, bins, beams and more. Wraps are designed for compatible Dymo handheld, industrial labelmakers (sold separately).
Catalog No. 50-007-39563 Supplier Enterprise Technology Solutions Supplier No. DYM1868705
Glossy wire/cable wraps feature a laminated overwrap to protect your printed areas from oil, chemicals, moisture, abrasion and fading. Thermal transfer printing technology protects from smearing and fading for long-lasting print visibility. Pre-sized design helps facilitate batch label printing to improve productivity. Wire/cable wraps are perfect for flat surface labeling, such as bar codes, faceplates, shelves, bins, beams and more. Wraps are designed for compatible Dymo handheld, industrial labelmakers (sold separately).