Learn More
Enterprise Technology Solutions Dymo DYMO D1 Electronic Tape, 1/2"x23' Size, Black/Clear
SDP
Catalog No. 5000740513
Encompass_Preferred
Created specifically for your LabelManager and LabelWriter Duo labelmakers, D1 labels offer the performance and variety you need for most labeling jobs. They adhere to most clean, flat surfaces, including plastic, metal and glass. Easy-peel backing means fast, trouble-free application. Versatile, durable D1 labels take the hassle out of home and office organization.
Catalog No. 50-007-40513 Supplier Enterprise Technology Solutions Supplier No. DYM45010
Created specifically for your LabelManager and LabelWriter Duo labelmakers, D1 labels offer the performance and variety you need for most labeling jobs. They adhere to most clean, flat surfaces, including plastic, metal and glass. Easy-peel backing means fast, trouble-free application. Versatile, durable D1 labels take the hassle out of home and office organization.