Learn More
Enterprise Technology Solutions Dymo Label, Flexible Nylon, 1/2"x11.5', Yellow
SDP
Catalog No. 5000740517
Encompass_Preferred
Flexible nylon label tape conforms and sticks tightly to wires, cables, curved and textured surfaces. Industrial-strength adhesive resists moisture, extreme temperatures, UV light and more - so labels won't curl or fall off. Using thermal transfer printing technology means the label text won't smudge, smear or fade. Easy-to-peel, split-back design makes applying labels quick and easy. Labels are compatible with Dymo Rhino 4200, 5200, 6000 and LW450 Duo.
Catalog No. 50-007-40517 Supplier Enterprise Technology Solutions Supplier No. DYM18490
Flexible nylon label tape conforms and sticks tightly to wires, cables, curved and textured surfaces. Industrial-strength adhesive resists moisture, extreme temperatures, UV light and more - so labels won't curl or fall off. Using thermal transfer printing technology means the label text won't smudge, smear or fade. Easy-to-peel, split-back design makes applying labels quick and easy. Labels are compatible with Dymo Rhino 4200, 5200, 6000 and LW450 Duo.