Learn More
Enterprise Technology Solutions Dymo Label, Flexible Nylon, 1"x11-1/2', White
SDP
Catalog No. 5000740643
Encompass_Preferred
1" flexible nylon label tape is specifically designed for wire and cable marking as well as other curved surfaces. Strong adhesion and flexibility make it ideal for industrial use, even where your label will encounter dirt and oil. 24mm labels are compatible with Dymo Rhino 6500, 6000, 5200.
Catalog No. 50-007-40643 Supplier Enterprise Technology Solutions Supplier No. DYM1734524
1" flexible nylon label tape is specifically designed for wire and cable marking as well as other curved surfaces. Strong adhesion and flexibility make it ideal for industrial use, even where your label will encounter dirt and oil. 24mm labels are compatible with Dymo Rhino 6500, 6000, 5200.