Learn More
Enterprise Technology Solutions Dymo Label, Heat Shrink Tube, 3/8"x5', White
SDP
Catalog No. 5000740634
Encompass_Preferred
White heat shrink tube offers a premium method of labeling wires and cables. Simply slide the tube onto the cable and shrink snugly to the cable using a heat gun. Labels will never fall off. The cassette fits into all Dymo Rhino printers including 6500, 6000, 5200, 5000, 4200, 3000 and 1000. Industrial-grade polyolefin offers a 3:1 heat shrink ratio and the convenience of printing directly on the 3/8" (9mm) tubing itself, eliminating the need for an extra labeling step.
Catalog No. 50-007-40634 Supplier Enterprise Technology Solutions Supplier No. DYM18053
White heat shrink tube offers a premium method of labeling wires and cables. Simply slide the tube onto the cable and shrink snugly to the cable using a heat gun. Labels will never fall off. The cassette fits into all Dymo Rhino printers including 6500, 6000, 5200, 5000, 4200, 3000 and 1000. Industrial-grade polyolefin offers a 3:1 heat shrink ratio and the convenience of printing directly on the 3/8" (9mm) tubing itself, eliminating the need for an extra labeling step.