Learn More
Enterprise Technology Solutions Dymo Label, Industrial, Perm Poly, 1/2"x18', White
SDP
Catalog No. 5000739644
Encompass_Preferred
Permanent poly labels are resistant to oil, solvents and chemicals. Durable tape adheres to all surfaces even extremely rough ones like name-and-rating plates, patch panels, cabinets and racks. Labels are compatible with Dymo Rhino 6500, 6000, 5200, 5000, 3000, and 1000.
Catalog No. 50-007-39644 Supplier Enterprise Technology Solutions Supplier No. DYM18483
Permanent poly labels are resistant to oil, solvents and chemicals. Durable tape adheres to all surfaces even extremely rough ones like name-and-rating plates, patch panels, cabinets and racks. Labels are compatible with Dymo Rhino 6500, 6000, 5200, 5000, 3000, and 1000.