Learn More
Enterprise Technology Solutions Dymo Label Tape,f/DYMO Labelmakers,1/2"x23',Black/White
SDP
Catalog No. 5000740655
Encompass_Preferred
Created specifically for your LabelManager and LabelWriter Duo labelmakers, D1 labels offer the performance and variety you need for most labeling jobs. They adhere to most clean, flat surfaces, including plastic, metal and glass. Easy-peel backing means fast, trouble-free application. Versatile, durable D1 labels take the hassle out of home and office organization. Labeling tape is 1/2" wide x 23' long with black print on white tape.
Catalog No. 50-007-40655 Supplier Enterprise Technology Solutions Supplier No. DYM45113
Created specifically for your LabelManager and LabelWriter Duo labelmakers, D1 labels offer the performance and variety you need for most labeling jobs. They adhere to most clean, flat surfaces, including plastic, metal and glass. Easy-peel backing means fast, trouble-free application. Versatile, durable D1 labels take the hassle out of home and office organization. Labeling tape is 1/2" wide x 23' long with black print on white tape.