Learn More
Enterprise Technology Solutions Dymo Label Tape,f/DYMO Labelmakers,3/8"x23',Black/Clear
SDP
Catalog No. 5000740652
Encompass_Preferred
Created specifically for your LabelManager and LabelWriter Duo labelmakers, D1 labels offer the performance and variety you need for most labeling jobs. They adhere to most clean, flat surfaces, including plastic, metal and glass. Easy-peel backing means fast, trouble-free application. Versatile, durable D1 labels take the hassle out of home and office organization. Labeling tape is 3/8" wide x 23' long with black print on clear tape.
Catalog No. 50-007-40652 Supplier Enterprise Technology Solutions Supplier No. DYMS0720670
Created specifically for your LabelManager and LabelWriter Duo labelmakers, D1 labels offer the performance and variety you need for most labeling jobs. They adhere to most clean, flat surfaces, including plastic, metal and glass. Easy-peel backing means fast, trouble-free application. Versatile, durable D1 labels take the hassle out of home and office organization. Labeling tape is 3/8" wide x 23' long with black print on clear tape.