Learn More
Enterprise Technology Solutions Dymo Labels,f/Labs/Test Tubes,1"x2-1/8",500/RL,WE
SDP
Catalog No. 5000739564
Encompass_Preferred
In a fast-paced laboratory or medical environment, you need labels that are reliable and withstand extreme temperatures, moisture, water, oil and chemicals. LW Durable Labels are ideal for test tube applications to reliably store samples and specimens in demanding environments. With 500 labels and 65 percent less waste thanks to Automatic Label Recognition, you can depend on getting the job done efficiently and with a high level of organization.
Catalog No. 50-007-39564 Supplier Enterprise Technology Solutions Supplier No. DYM2187918
In a fast-paced laboratory or medical environment, you need labels that are reliable and withstand extreme temperatures, moisture, water, oil and chemicals. LW Durable Labels are ideal for test tube applications to reliably store samples and specimens in demanding environments. With 500 labels and 65 percent less waste thanks to Automatic Label Recognition, you can depend on getting the job done efficiently and with a high level of organization.