Learn More
GA International DYMO LabelWriter 550 Direct Thermal Printer
Catalog No. 5000819164
Encompass_Preferred
The DYMO LabelWriter 550 is a direct thermal printer that does not require costly inks or toner to print high-quality text and barcodes, while its compact design allows it to fit seamlessly into any workspace. Its patented automatic label recognition technology will automatically identify label size and type, allowing it to quickly generate the template once you load the labels, so you never have to waste time selecting the right template. In addition, the automatic label counter indicates how many labels remain—so you’ll never run out unexpectedly. Free DYMO Connect software is included with every printer, allowing you to create 1D and 2D barcodes along with crisp, clear text and graphics. The 550 is up to 20% faster than its predecessor, the LabelWriter 450, and is fully compatible with our LWCS-class of cryogenic labels.
Catalog No. 50-008-19164 Supplier GA International Supplier No. DYMOLW550NA
The DYMO LabelWriter 550 is a direct thermal printer that does not require costly inks or toner to print high-quality text and barcodes, while its compact design allows it to fit seamlessly into any workspace. Its patented automatic label recognition technology will automatically identify label size and type, allowing it to quickly generate the template once you load the labels, so you never have to waste time selecting the right template. In addition, the automatic label counter indicates how many labels remain—so you’ll never run out unexpectedly. Free DYMO Connect software is included with every printer, allowing you to create 1D and 2D barcodes along with crisp, clear text and graphics. The 550 is up to 20% faster than its predecessor, the LabelWriter 450, and is fully compatible with our LWCS-class of cryogenic labels.