Learn More
MSC Dymo Permanent Polyester Tape, Clear, 3/8 x 18, 1 Roll
Catalog No. 5000769946
Encompass
This permanent polyester tape is designed for reliable and durable labeling applications. It is compatible with Dymo label makers, including the 5000, 5200, 6000, 6500, and Rhino 3000 models.
  • Permanent polyester tape for long-lasting adhesion
  • Clear color for professional appearance
  • Compatible with Dymo 5000, 5200, 6000, 6500, and Rhino 3000 label makers
  • Width of 3/8 in
  • Length of 18 ft
  • Comes as 1 roll
Catalog No. 50-007-69946 Supplier MSC Supplier No. 07591365
This permanent polyester tape is designed for reliable and durable labeling applications. It is compatible with Dymo label makers, including the 5000, 5200, 6000, 6500, and Rhino 3000 models.
  • Permanent polyester tape for long-lasting adhesion
  • Clear color for professional appearance
  • Compatible with Dymo 5000, 5200, 6000, 6500, and Rhino 3000 label makers
  • Width of 3/8 in
  • Length of 18 ft
  • Comes as 1 roll