Learn More
Enterprise Technology Solutions Dymo Printhead Cleaning Cards,f/LabelWriter,10/BX,WE
SDP
Catalog No. 5000739638
Encompass_Preferred
Printhead Cleaning Cards safely and effectively remove contamination from the printhead, platen and label path of your Dymo LabelWriter. Use these cards once or twice a month or whenever print quality diminishes. Cleaning cards improve print quality and extend the life of your LabelWriter.
Catalog No. 50-007-39638 Supplier Enterprise Technology Solutions Supplier No. DYM60622
Printhead Cleaning Cards safely and effectively remove contamination from the printhead, platen and label path of your Dymo LabelWriter. Use these cards once or twice a month or whenever print quality diminishes. Cleaning cards improve print quality and extend the life of your LabelWriter.