Learn More
MSC Dymo RHINO 5200 Labeling Tool Kit, 180 DPI, 1 Kit
Catalog No. 5000769780
Encompass
The Professional Handheld Labeling Tool Kit is a battery-powered device designed for professional labeling applications, featuring a resolution of 180 dpi.
  • Includes RHINO 5200 printer
  • Comes with rugged carrying case
  • Powered by rechargeable lithium-ion battery pack
  • Includes AC adapter for charging
  • Contains 3/4 in white flexible nylon tape
  • Contains 3/8 in white vinyl tape
  • Provided with quick reference guide
  • User manual on CD Rom included
Catalog No. 50-007-69780 Supplier MSC Supplier No. 07591357
The Professional Handheld Labeling Tool Kit is a battery-powered device designed for professional labeling applications, featuring a resolution of 180 dpi.
  • Includes RHINO 5200 printer
  • Comes with rugged carrying case
  • Powered by rechargeable lithium-ion battery pack
  • Includes AC adapter for charging
  • Contains 3/4 in white flexible nylon tape
  • Contains 3/8 in white vinyl tape
  • Provided with quick reference guide
  • User manual on CD Rom included