Learn More
Enterprise Technology Solutions Dymo Shipping Labels, Polyester, 3/4"x18', White
SDP
Catalog No. 5000739554
Encompass_Preferred
Permanent polyester tape features durable construction designed for industrial use. 3/4" (19mm) poly label tape is resistant to oil, solvents and chemicals and adheres to all surfaces, even extremely rough ones like name-and-rating plates, patch panels, cabinets and racks. The split back on the adhesive side makes the tape easy to peel. The permanent adhesive results in labels that stick and last in rugged environments. Tape is compatible with Dymo Rhino 6500, 6000, 5200 and 5000.
Catalog No. 50-007-39554 Supplier Enterprise Technology Solutions Supplier No. DYM18484
Permanent polyester tape features durable construction designed for industrial use. 3/4" (19mm) poly label tape is resistant to oil, solvents and chemicals and adheres to all surfaces, even extremely rough ones like name-and-rating plates, patch panels, cabinets and racks. The split back on the adhesive side makes the tape easy to peel. The permanent adhesive results in labels that stick and last in rugged environments. Tape is compatible with Dymo Rhino 6500, 6000, 5200 and 5000.