Learn More
Enterprise Technology Solutions Dymo Vinyl Labels, 3/4"x18ft, Black/Yellow
SDP
Catalog No. 5000739722
Encompass_Preferred
Colored vinyl labels are a perfect solution for general labeling in and around a facility or where colored labels are required. Color-coding with black text gives a better visual recognition to your identification needs. These 3/4" labels for industrial purposes are ideal for warning and safety messages, patch panels/faceplates and general labeling where color labels are required. Additionally, vinyl labels can be used outdoors in direct sunlight. Labels are compatible with Dymo Rhino 6500, 6000, 5200 and 5000.
Catalog No. 50-007-39722 Supplier Enterprise Technology Solutions Supplier No. DYM18433
Colored vinyl labels are a perfect solution for general labeling in and around a facility or where colored labels are required. Color-coding with black text gives a better visual recognition to your identification needs. These 3/4" labels for industrial purposes are ideal for warning and safety messages, patch panels/faceplates and general labeling where color labels are required. Additionally, vinyl labels can be used outdoors in direct sunlight. Labels are compatible with Dymo Rhino 6500, 6000, 5200 and 5000.