Learn More
Enterprise Technology Solutions Dymo XTL All-Purpose Vinyl Labels, 1", BKWE
SDP
Catalog No. 5000739767
Encompass_Preferred
XTL All-purpose Vinyl Labels are durable and ideal for almost any labeling application from textured surfaces to cables. Whether you are labeling to OSHA, ISO and ANSI color standards or labeling in harsh conditions, these labels have you covered. Jobsite-tested, UL-recognized labels won't smear or fade. Use with Dymo XTL labelmakers. Vinyl offers moisture-resistance, chemical-resistance and UV-resistance, making these black-on-white labels ideal for labeling both indoors and outdoors. Easy-to-peel, split-back design makes applying labels quick and easy.
Catalog No. 50-007-39767 Supplier Enterprise Technology Solutions Supplier No. DYM1868753
XTL All-purpose Vinyl Labels are durable and ideal for almost any labeling application from textured surfaces to cables. Whether you are labeling to OSHA, ISO and ANSI color standards or labeling in harsh conditions, these labels have you covered. Jobsite-tested, UL-recognized labels won't smear or fade. Use with Dymo XTL labelmakers. Vinyl offers moisture-resistance, chemical-resistance and UV-resistance, making these black-on-white labels ideal for labeling both indoors and outdoors. Easy-to-peel, split-back design makes applying labels quick and easy.