Learn More
MSC Dyna-Glo LP Tank Top Heater, 15000 BTU, 1 ea
Catalog No. 5000502626
Encompass
The Dyna-Glo TT15CDGP 15,000 BTU LP Tank Top Heater is a CSA certified radiant propane heater designed for temporary heating applications. It features a variable supply valve, offering outputs from 9,000 up to 15,000 BTUs.
  • Variable BTU output (9,000 to 15,000 BTUs)
  • CSA certified safety tip switch
  • Quick connect P.O.L. tank connection
  • Safety shut-off valve
  • CSA certified high wind resistant burner
  • Portable and stable design
  • Match ignition system
Catalog No. 50-005-02626 Supplier MSC Supplier No. 53772414
The Dyna-Glo TT15CDGP 15,000 BTU LP Tank Top Heater is a CSA certified radiant propane heater designed for temporary heating applications. It features a variable supply valve, offering outputs from 9,000 up to 15,000 BTUs.
  • Variable BTU output (9,000 to 15,000 BTUs)
  • CSA certified safety tip switch
  • Quick connect P.O.L. tank connection
  • Safety shut-off valve
  • CSA certified high wind resistant burner
  • Portable and stable design
  • Match ignition system