Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ Biotin Binder

Catalog No. p-200105789
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-048-D 2 mL
11-047 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-048-D Supplier Invitrogen™ Supplier No. 11048D
Only null left
Add to Cart
Edge
Add to Cart

Dynabeads Biotin Binder contains 2.8 μm superparamagnetic beads that bind biotinylated antibodies (or other biotinylated ligands), allowing you to isolate cells from any species (e.g., human, mouse, rat, non-human primates, and cell lines).

Dynabeads Biotin Binder contains 2.8 μm superparamagnetic beads that bind biotinylated antibodies (or other biotinylated ligands), allowing you to isolate cells from any species (e.g., human, mouse, rat, non-human primates, and cell lines). Advantages of Dynabeads Biotin Binder:

  • Works with any cell type, from any sample and from any species
  • Fast and efficient depletion or positive isolation of any target cells for molecular downstream assays using a biotinylated primary antibody
  • Obtain pure and untouched cells using a mixture of biotinylated antibodies to deplete unwanted cells

Adaptable for various isolation strategies

The biotinylated primary antibody may be added to the cell sample (indirect technique) or pre-coated onto the Dynabeads Biotin Binder (direct technique). Regardless of which technique is used, when the cell sample and the Dynabeads Biotin Binder are mixed, the beads bind to the target cells. Placing the sample on a magnet separates the bead-bound cells from the rest of the sample. One or more biotinylated antibodies may be used to either deplete the sample of unwanted cells (negative isolation) or to capture specific cells for downstream experiments (positive isolation). For isolation of bead-free cells (e.g., for flow cytometry), we recommend using your biotinylated antibody in combination with the CELLection Biotin Binder Kit.

Useful for many sample types

Using Dynabeads Biotin Binder, you can isolate cells basically from any species, and from many starting samples such as whole blood, cord blood, peripheral blood mononuclear cells (PBMC), buffy coat, mouse spleen/lymph nodes, and tissue digests.

Learn more about Dynabeads products

TRUSTED_SUSTAINABILITY

Specifications

Cell Type All cells from all species
Content And Storage This Product Contains: 2 mL Dynabeads™ coated with recombinant streptavidin.
Color Brown
For Use With (Application) Cell Depletion, Negative Isolation, Positive Isolation
Material Magnetic Polystyrene
Product Type Biotin Binder
No. of Cells Processes ∼8x 10^8 cells total
Output Viability >95%
Product Line Dynabeads
Purity or Quality Grade Research Grade
Quantity 2 mL
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 10^7 PBMCs per isolation
Target Species All species
Diameter (Metric) 2.8 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.