Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Dynabeads™ Human T-Expander CD3/CD28

Catalog No. 11141D
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-141-D 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-141-D Supplier Gibco™ Supplier No. 11141D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dynabeads™ Human T-Expander CD3/CD28

Dynabeads™ Human T-Expander CD3/CD28 is intended for separation, activation and expansion of human T cells. Dynabeads™ Human T-Expander CD3/CD28 is the research grade version of Dynabeads™ CD3/CD28 CTS™. Containing the same amount and ratio of antibodies from the same clones as Dynabeads™ CD3/CD28 CTS™, Dynabeads™ Human T-Expander CD3/CD28 are intended for small scale pre-clinical research. Dynabeads™ Human T-Expander CD3/CD28 offers a simple method for activation and expansion of T-cells that does not require antigen presenting cells or antigen. By combining anti-CD3 and anti-CD28 antibodies on Dynabeads™, the beads provide both the primary and co-stimulatory signals that are required for activation and expansion of T cells.

Applications:
Dynabeads™ Human T-Expander CD3/CD28 is the research grade version of Dynabeads™ CD3/CD28 CTS™. The Dynabeads™ CD3/CD28 CTS™ technology has been successfully used in ex vivo separation, activation and expansion of T-cells in a number of studies, including studies on HIV infection and cancer. Other potential applications for this novel technology are treatment of autoimmune and infectious diseases and prevention of complications associated with chemotherapy and allogeneic transplantation.

Specifications

Cell Type T Cells (Whole Population)
Clonality Monoclonal
Content And Storage This Product Contains: Dynabeads™ CD3⁄CD28 is supplied as a suspension containing 1 x 108 beads⁄ml in phosphate buffered saline (PBS), pH 7.4, wwith 0.1% human serum albumin (HSA).
Store at: 2-8°C
Product Type Antibody Coated Bead
Final Product Type Cells
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 10 mL
Reactivity Human
Sample Type Cell Cultures, PBMC
Shipping Condition Room Temperature
Target Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.