Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ Streptavidin for In Vitro Transcription

Catalog No. p-7229379
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
65-005-D 2 mL
65-006-D 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-005-D Supplier Invitrogen™ Supplier No. 65005D
Only null left
Add to Cart
Edge
Add to Cart

Dynabeads Streptavidin for In Vitro Transcription offers an innovative technology for simple, flexible, and automation-ready mRNA synthesis in scales ranging from microgram to gram.

Dynabeads Streptavidin for In Vitro Transcription offer an innovative technology for simple, flexible, and automatable mRNA synthesis, in scales ranging from micrograms to grams. They are particularly suitable where flexibility and high quality are required, such as in the discovery phase of RNA-based vaccines and therapeutics development.

The 1-μm monosized, monodispersed, paramagnetic beads are covalently coupled with streptavidin molecules, for binding to biotinylated DNA template. The workflow is designed to minimize the effort and cost of plasmid preparation, while enabling reuse of template-bead complex in successive solid-phase in vitro transcription (IVT) reactions (figure below).

Features of Dynabeads Streptavidin for In Vitro Transcription include:

  • Flexibility – choose DNA template for IVT with flexibility of design and amplification
  • Efficiency – reuse DNA template and beads at least six times to help reduce cost and eliminate/minimize plasmid prep hands-on time and risk of bacterial host DNA transfer
  • Scalability – swap between manual and automated workflows to match required scale (micrograms to grams of mRNA) and throughput
  • Quality – help ensure a safe and flexible supply of magnetic beads to navigate from RNA-based drug discovery research to commercial manufacturing with counterpart solutions (available via the OEM supply) without incurring extra time or costs from switching technology or raw material

Overview of the workflow
A biotinylated template is made by PCR amplification of the target sequence in a plasmid construct or in a synthetic DNA construct using a biotinylated forward primer and a non-biotinylated reverse primer. As an alternative to PCR amplification, a linearized plasmid can be biotinylated by a fill-in reaction of biotin-dUTP in the 5' overhang sequence, provided the correct plasmid design. The biotinylated template is then immobilized directly to the Dynabeads Streptavidin for In Vitro Transcription beads without the need for prior purification. The bead-bound template is used directly in the IVT, then removed from the synthesized mRNA by magnetic separation (Figure 1). The template can be reused (at least six times) in consecutive IVT reactions (figure below).

Solid-phase IVT is compatible with commonly used reagents such as MEGAscript T7 Transcription Kit, mMESSAGE mMACHINE T7 Transcription Kit, and mMESSAGE mMACHINE T7 ULTRA Transcription Kit, based on your needs and strategies for capping and polyadenylation.

The beads are offered here as a stand-alone product. They are also included in the Dynabeads In Vitro Transcription and RNA Purification Kit.

Automation-ready for use with KingFisher purification systems
The 1-μm magnetic beads are ideal for high-throughput screening. The process can be automated using liquid handlers or any of our KingFisher sample purification systems. The scripts for KingFisher Flex and KingFisher Apex automated purification workflows are available from our website.

Commercial supply
To learn more about counterpart products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics, visit: thermofisher.com/dynabeadsmrna

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mg/mL
Color Brown
Content And Storage Dynabeads Streptavidin for In Vitro Transcription is supplied in PBS, pH 7.4, with 20% ethanol and 0.01% Tween-20. Store at 2°C to 8°C.
Diameter (Metric) 1 μm
Isolation Technology Magnetic Bead
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
For Use With (Application) RNA Purification
Product Line Dynabeads
Product Type Streptavidin Magnetic Beads
Quantity 2 mL
Shipping Condition Ambient Temperature
Volume (Metric) 2 mL
Ligand Type Biotin
Certifications/Compliance ISO9001 and ISO13485
Material Magnetic Polystyrene
Uniformity Monosized 1 μm (CV <5%)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.