Learn More
Ace Glass, Inc. Dynabloc segmented block, 12 (13x27.6mm) holes for 1/2 dram vials, 12 (15x27.6mm) holes for 1 dram vials, 8 (17x27.6mm)holes for 2 dram vials
SDP
Catalog No. 50978102
Encompass
Anodized aluminum Dynabloc heating/mixing blocks with three segments to hold up to three different size and type of vessels at the same time. Dynabloc has 12 (13x27.6mm) holes for dram vials, 12 (15x27.6mm) holes for 1 dram vials, and 8 (17x27.6mm) holes for 2 dram vials. Dynablocs should be anchored to either a 135mm (50978108) or 145mm (50983541) base plate. A T-handle extractor (50978109) should be used to remove blocks when hot.
Catalog No. 50-978-102 Supplier Ace Glass, Inc. Supplier No. 1369603
Anodized aluminum Dynabloc heating/mixing blocks with three segments to hold up to three different size and type of vessels at the same time. Dynabloc has 12 (13x27.6mm) holes for dram vials, 12 (15x27.6mm) holes for 1 dram vials, and 8 (17x27.6mm) holes for 2 dram vials. Dynablocs should be anchored to either a 135mm (50978108) or 145mm (50983541) base plate. A T-handle extractor (50978109) should be used to remove blocks when hot.