Learn More
Ace Glass, Inc. Dynabloc block, 1/2 dram 2mL vial, 54 (13x27.6mm) holes
SDP
Catalog No. 50983544
Encompass
Dram, 12mL vial size Dynabloc block with 54 size 13x27.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.
Catalog No. 50-983-544 Supplier Ace Glass, Inc. Supplier No. 1369849
Dram, 12mL vial size Dynabloc block with 54 size 13x27.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.