Learn More
Ace Glass, Inc. Dynabloc segmented block, 12 (15x27.6mm) holes for 1 dram vials, 8 (17x27.6mm) holes for 2 dram vials, 4 (22x27.6mm)holes for 4 dram vials
SDP
Catalog No. 50978103
Encompass
Anodized aluminum Dynabloc heating/mixing blocks with three segments to hold up to three different size and type of vessels at the same time. Dynabloc has 12 (15x27.6mm) holes for 1 dram vials, 8 (17x27.6mm) holes for 2 dram vials, and 4 (22x27.6mm) holes for 4 dram vials. Dynablocs should be anchored to either a 135mm (50978108) or 145mm (50983541) base plate. A T-handle extractor (50978109) should be used to remove blocks when hot.
Catalog No. 50-978-103 Supplier Ace Glass, Inc. Supplier No. 1369607
Anodized aluminum Dynabloc heating/mixing blocks with three segments to hold up to three different size and type of vessels at the same time. Dynabloc has 12 (15x27.6mm) holes for 1 dram vials, 8 (17x27.6mm) holes for 2 dram vials, and 4 (22x27.6mm) holes for 4 dram vials. Dynablocs should be anchored to either a 135mm (50978108) or 145mm (50983541) base plate. A T-handle extractor (50978109) should be used to remove blocks when hot.