Learn More
Ace Glass, Inc. Dynabloc block, 1mL V-vial size, 28 (13x27.6mm) holes
SDP
Catalog No. 50978113
Encompass
1mL V-vial size Dynabloc block with 28 size 13x27.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.
Catalog No. 50-978-113 Supplier Ace Glass, Inc. Supplier No. 1369821
1mL V-vial size Dynabloc block with 28 size 13x27.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.