Learn More
Ace Glass, Inc. Dynabloc block, 20mL & 30mL vial size, 18 (28x27.6mm) holes
SDP
Catalog No. 50978110
Encompass
20mL & 30mL vial size Dynabloc block with 18 size 28x27.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.
Catalog No. 50-978-110 Supplier Ace Glass, Inc. Supplier No. 1369812
20mL & 30mL vial size Dynabloc block with 18 size 28x27.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.