Learn More
Ace Glass, Inc. Dynabloc block, 4 dram 16mL vial size, 28 (22x35.6mm) holes
SDP
Catalog No. 50978112
Encompass
4 dram, 16mL vial size Dynabloc block with 28 size 22x35.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.
Catalog No. 50-978-112 Supplier Ace Glass, Inc. Supplier No. 1369820
4 dram, 16mL vial size Dynabloc block with 28 size 22x35.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.