Learn More
Ace Glass, Inc. Dynabloc block, 40mL vial size, 18 (28x50mm) holes
SDP
Catalog No. 50983542
Encompass
40mL vial size Dynabloc block with 18 size 28x50mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.
Catalog No. 50-983-542 Supplier Ace Glass, Inc. Supplier No. 1369813
40mL vial size Dynabloc block with 18 size 28x50mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.