Learn More
Ace Glass, Inc. DYNABLOC EXTRACTOR DYNABLOC EXTRACTOR
SDP
Catalog No. 50978109
Encompass
Dynabloc T-handle extractor for removing blocks when hot.
Catalog No. 50-978-109 Supplier Ace Glass, Inc. Supplier No. 1369809
Dynabloc T-handle extractor for removing blocks when hot.