Learn More
Ace Glass, Inc. Dynabloc, heating block, flask, 125mL Erlenmeyer size, (3) 69mmwells
SDP
Catalog No. 50978117
Encompass
DynaBloc heating block with (3) 69mm wells, for use with 125mL Erlenmeyer flasks. Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/or heating of multiple samples contained within small, round bottom flasks. When hot, blocks should be removed using a T-handle DynaBloc extractor (Part 50978109).
Catalog No. 50-978-117 Supplier Ace Glass, Inc. Supplier No. 1369903
DynaBloc heating block with (3) 69mm wells, for use with 125mL Erlenmeyer flasks. Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/or heating of multiple samples contained within small, round bottom flasks. When hot, blocks should be removed using a T-handle DynaBloc extractor (Part 50978109).