Learn More
Thomas Scientific Plate, 96 x 1.0 mL/Round Well, Polypropylene, Sterile
SDP
Catalog No. 501458613
Encompass_Preferred
These deep well microplates are ideal for high throughput applications and storage of samples. They are compatible with Labnet’s AccuSeal Plate Sealer, sample processing workstations, microplate processing equipment and many microplate centrifuge rotors. Ideal for high throughput applications and storage of samples DNase/RNase free, sterile and non-sterile packaging Steel freezer storage racks available Bar coding available upon request Plates with 0.5 and 1.0 mL well volumes have round wells while 2.0 mL plates have square wells. All wells have conical bottoms to aid in sample recovery. All of the plates are available in polypropylene for excellent temperature and chemical resistance. They can be frozen or autoclaved, and when used with sealing mats (available separately), are perfect for freezer storage of samples. Sealing mats are available for the 96 well plates. The mats seal each well individually. Plates are DNAse/RNase free and supplied sterile or nonsterile.
Catalog No. 50-145-8613 Supplier Thomas Scientific Supplier No. 1201T72
These deep well microplates are ideal for high throughput applications and storage of samples. They are compatible with Labnet’s AccuSeal Plate Sealer, sample processing workstations, microplate processing equipment and many microplate centrifuge rotors. Ideal for high throughput applications and storage of samples DNase/RNase free, sterile and non-sterile packaging Steel freezer storage racks available Bar coding available upon request Plates with 0.5 and 1.0 mL well volumes have round wells while 2.0 mL plates have square wells. All wells have conical bottoms to aid in sample recovery. All of the plates are available in polypropylene for excellent temperature and chemical resistance. They can be frozen or autoclaved, and when used with sealing mats (available separately), are perfect for freezer storage of samples. Sealing mats are available for the 96 well plates. The mats seal each well individually. Plates are DNAse/RNase free and supplied sterile or nonsterile.