Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ M-280 Tosylactivated

Catalog No. 142040
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-204-0 10 mL
14-203 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-204-0 Supplier Invitrogen™ Supplier No. 14204
  • $1,858.00
    $1,634.65 / Each of 1

    Save $223.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

When you wish to covalently couple antibodies, peptides, intact proteins and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated.

When you wish to covalently couple antibodies, peptides, intact proteins and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated. Optimal antibody orientation makes these beads an excellent choice for the immunoprecipitation of proteins and protein complexes.

Ideal for Immunoprecipitation of protein complexes:
Low background and covalent linking of antibodies to the bead surface make Dynabeads Tosylactivated an excellent choice for Immunoprecipitation of proteins and protein complexes (Co-Immunoprecipitation, Co-IP). Gentle and rapid magnetic concentration of the beads and short incubation times (possible due to fast surface-based binding kinetics) make Dynabeads Tosylactivated an excellent choice for the immunoprecipitation of highly-labile or and/or transient (short-lived) protein complexes.

Ultra rapid protocols:

  • Identify members of protein complexes in minutes rather than hours
  • Temporal resolution short enough to identify transient and labile complexes
  • Identify binding partners that cannot be identified with longer protocols
  • Rapid protocols further reduce the already ultra-low background binding
  • Stronger signal to noise-ratios

Applications:

  • IP of proteins and protein complexes
  • Couple functional enzymes to bead surface for downstream assays
  • Couple peptides to bead surface to identify binders

Purify peptides, proteins and enzymes that are:

  • Transiently stable
  • Structurally intact
  • Temperature labile
  • In their native conformation and functional

Coupling Procedure Outline:
Covalent coupling is performed overnight by incubating the desired ligand with the Dynabeads Tosylactivated. Ligands commonly coupled to Dynabeads Tosylactivated include peptides and proteins (e.g. antibodies for Immunoprecipitation or Co-Immunoprecipitation). Coupling occurs at neutral to high pH and at 37°C. We recommend coupling at pH 8.5–9.5, but for pH labile ligands, coupling can be performed in an alternative buffer at pH 7.4.

Upon completion of the ligand coupling step, the actual Dynabeads Tosylactivated surface coating will be rendered inert, resulting in low non-specific binding.

About Dynabeads:
Dynabeads are non-porous monodisperse superparamagnetic beads. They are highly mobile in solution enabling ligands coupled to the beads to continuously interact with the entire sample volume. The superparamagnetic beads are pulled to the tube walls by transferring the tube to a rack containing a strong magnetic field (From the red navigation bar navigate Brands > Dynal > Magnets). Strong magnetic fields quickly pull the beads to the tube wall, allowing for easy and complete removal of the supernatant by pipette. Washing steps are performed similarly.l

Bead Surface Characteristics for Dynabeads Tosylactivated:

  • p-toluene-sulfonyl (Tosyl) groups
  • Hydrophobic, pH Neutral
  • Covalent binding by primary amine (NH2) or sulphydryl (SH) groups

Binding capacity per milligram beads:
Varies depending on ligand, (e.g. 5–10 μg IgG)

Order Info

Shipping Condition: Room Temperature

Warranty and Services

General Terms and Conditions

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C.
Elution Volume Min 5 μL
High-throughput Compatibility High-throughput Compatible
Shipping Condition Room Temperature
Sample Type Any Sample Type
Surface Functionality p-Toluene-Sulfonyl, Tosylactivated
Product Line DYNAL, Dynabeads
Quantity 10 mL
Type Tosylactivated Bead
Format Beads in suspension
Purity or Quality Grade Research Grade
Show More Show Less
My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

What sizes do you offer for the Dynabeads magnetic beads?

Dynabeads magnetic beads come in three sizes: 4.5 µm (M-450), 2.8 µm (M-270/M-280), and 1 µm (MyOne beads). The largest of the Dynabeads magnetic beads is ideal for big targets like cells. The 2.8 µm beads are recommended for proteomics and molecular applications. The smallest of the beads, 1 µm, are ideal for automated handling.

Can Dynabeads magnetic beads be sonicated?

In general, short sonication is a good way to reduce aggregation of the beads and ensure optimal homogenous conditions at the time of ligand addition when coating the beads. When target is bound to the beads, more care is needed, as the binding might break. The streptavidin beads themselves should tolerate sonication. We have not tested sonication for long periods, but 5 minutes is fine. We do not have information about the streptavidin-biotin interaction being broken by such treatment.

Can Dynabeads magnetic beads be sterilized?

If desired, the uncoated epoxy or tosylactivated beads can be sterilized by washing with 70% ethanol. Coated beads cannot be sterilized.

What reactive groups do you offer for your surface-activated Dynabeads magnetic beads?

We offer tosyl, epoxy, carboxylic acid and amine activated Dynabeads magnetic beads. Please see the link (https://tools.thermofisher.com/content/sfs/brochures/Surface_Activated_Dynabeads.PDF) for a comparison of the beads.

What are Dynabeads magnetic beads?

Dynabeads magnetic beads are uniform, non-porous, superparamagnetic, monodispersed and highly cross-linked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The magnetic material within the Dynabeads magnetic beads consists of a mixture of maghemite (gamma-Fe2O3) and magnetite (Fe3O4). The iron content (Fe) of the beads is 12% by weight in Dynabeads magnetic beads M-280 and 20% by weight in Dynabeads magnetic beads M-450. The Dynabeads magnetic beads are coated with a thin polystyrene shell which encases the magnetic material, and prevents any leakage from the beads or trapping of ligands in the bead interior. The shell also protects the target from exposure to iron while providing a defined surface area for the adsorption or coupling of various molecules.

Uniformity of bead size and shape provides consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.

The Dynabeads magnetic beads are available in three different sizes: 4.5 µm (M-450 beads), 2.8 µm (M-270/M-280 beads) and 1 µm (MyOne beads). as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

For how long can Dynabeads magnetic beads be stored?

Storage should be at 2-8 degrees C. Freezing Dynabeads magnetic beads is not recommended. Provided the Dynabeads magnetic beads are stored correctly, quality is guaranteed until the expiration date stated on the label. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How should I store coated Dynabeads magnetic beads if I want to reuse them?

Dynabeads magnetic beads coated with antibody/ligand may be stored at 2-8 degrees C without loss of antigen binding capacity. For long-term storage, a final concentration of 0.02% NaN3 may be added to the antibody-coupled beads in a physiological buffer. Please note that not all coupled antibodies retain their function in long term storage. Verify your coupled antibody stability by testing in small scale. After storage, coated Dynabeads magnetic beads should be washed once in PBS/BSA for 5 min before use.

How can I dissociate isolated proteins from Dynabeads magnetic beads?

Standard elution methods are used to dissociate the isolated protein from Dynabeads magnetic beads. The most suitable elution method depends on the characteristics of the isolated protein and the desired downstream application of the eluted protein. In many cases, Dynabeads magnetic beads can be recovered for reuse after elution. Some standard elution methods are listed below.

-pH change: elution can be achieved by reducing pH (for example, by using 0.1 M citrate (pH 2-3) as the elution buffer). Dynabeads magnetic beads are stable between pH 4-13. If the Dynabeads magnetic beads are exposed for a prolonged period of time to pH below 4, the beads may be adversely affected.

-Change of ionic strength: high-salt concentration buffers (e.g., NaI, KI, MgCl, KCl) can be used to elute isolated proteins. Optimization is required, by step-wise elution starting at 1 M and increasing to 3 M.

-Affinity elution: with this method, the eluting agent competes for the binding of the protein or the binding of the ligand e.g., elution of glycoproteins from a lectin coupled to Dynabeads magnetic beads may be achieved by the addition of the free sugar.

-Denaturing eluents: as a last resort, denaturing eluents such as chaotropic salts may be used to alter the structure of the protein. The proteins on the bead surface and the eluted proteins will be denatured.

-Polarity reducing agents : substances that reduce the polarity of the buffer often disrupt the hydrophobic interactions between antibody and protein. Dioxane or ethylene glycol may be used to reduce the polarity of the eluent. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the definition of superparamagnetic, and what does this mean for my cell isolation application with Dynabeads magnetic beads?

Superparamagnetic means that the Dynabeads magnetic beads exhibit magnetic properties when placed within a magnetic field, but have no residual magnetism when removed from the magnetic field.

This means that your targeted cells, proteins, or nucleic acids are only subjected to magnetic forces during the time the beads are on the magnet. The beads do not aggregate, but remain evenly dispersed in suspension.

Are the antibodies on your Dynabeads magnetic beads for cell isolation/activation/expansion covalently bound to the beads?

Yes. The antibodies are covalently bound and should be very stable in your applications.

What is the average shelf life of Dynabeads magnetic beads?

Depending on the antibody coated on the Dynabeads magnetic beads, the shelf life can vary from 24-36 months.
Some kits may have 18 months shelf life depending on other components supplied in the kit. The kits are guaranteed for 6 months from when you receive them. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Can I store the antibody/ligand-coated Dynabeads magnetic beads during isolation of cell organelles?

Dynabeads magnetic beads coated with antibody/ligand may be stored at 2-8 degrees C for several weeks without loss of antigen binding capacity. After storage, coated Dynabeads magnetic beads should be washed once in PBS/BSA for 5 mins before use.

Which surface-activated Dynabeads magnetic beads should I choose for immobilizing my protein?

This depends on the nature of the specific ligand to be immobilized and the desired downstream application.

-The most frequently used surface-activated Dynabeads magnetic beads for protein isolation are the Tosyl-activated Dynabeads magnetic beads M-280. These beads are hydrophobic, easy to handle and ideal for covalent binding of antibodies for immunoprecipitation. Other ligands could also be covalently bound to these beads, but they have to tolerate conditions like neutral to high pH and high temperatures (required for covalent bond formation).
-Dynabeads magnetic beads M-270 Epoxy are used when the ligand to be immobilized needs to be treated gently and will not tolerate harsh binding conditions like high temperature or pH. Proteins/peptides (other than antibodies) and enzymes are often coupled onto these beads.
-Dynabeads magnetic beads M-270 Amine is often used in combination with crosslinkers to create specific surface groups on the beads. Hetro-bifunctional crosslinkers with an amine-reactive NHS group at one end and another chemical group of your choice at the other end are most frequently used. For example, Dynabeads magnetic beads M-270 Amine can be reacted with a hetero-bifunctional crosslinker containing a NHS group at one end and maleimide at the other end to create Dynabeads magnetic beads with a maleimide surface. Since maleimide reacts specifically with sulfhydryl groups, these modified Dynabeads magnetic beads can be used for applications where binding of sulfhydryl groups are desired. Dynabeads magnetic beads M-270 Amine may also be used for direct ligand coupling via aldehyde or ketone groups by Schiff base (imine) formation and reductive amination. In addition carboxylic acid groups on a ligand can be activated with a carbodiimide like EDC, which results in a direct amide bond formation between the beads and the ligand. Alternatively, a crosslinker may be introduced to the ligand. After activation of the ligand with crosslinker, the free end on the crosslinker has to be amine reactive.
-Dynabeads magnetic beads M-270 Carboxylic Acid and Dynabeads magnetic beads MyOne Carboxylic Acid can also be used for immobilizing proteins. The carboxylic acid groups on these beads need to be activated with a carbodiimide before coupling. The negatively-charged surface of these beads may attract positively charged proteins and cause nonspecific binding. This needs to be considered if these beads are going to be used for immunoprecipitation.

How are proteins coupled to surface-activated Dynabeads magnetic beads?

Dynabeads M-280 Tosyl activated magnetic beads allow direct covalent binding to primary amino or sulfhydryl groups in proteins and peptides at high pH and high temperature.
-Dynabeads M-270 Epoxy magnetic beads allow direct covalent binding to primary amino and sulfhydryl groups in proteins and peptides at neutral pH and across a wide temperature range.
-Dynabeads M-270 Amine magnetic beads allow direct covalent binding through reductive amination of aldehydes, or the use of bifunctional amine-reactive crosslinkers.
-Dynabeads M-270 and MyOne Carboxylic Acid magnetic beads allow covalent amide formation with primary amino groups in proteins and peptides.

What do the designations M-280, M-270, and MyOne mean on Dynabeads magnetic beads?

The M stands for magnetic. M-280 refers to hydrophobic 2.8 micron beads, while M-270 refers to hydrophilic 2.8 micron beads. MyOne refers to 1 micron beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the detection limit when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Answering this question is not straightforward. It will depend on the detection method. When using HRP (horseradish peroxidase)-based detection system or radioactivity in combination with a good antibody, very little target is required. More target is required when using an AP (alkaline phosphatase)-based detection system. When a sensitive detection system is used, detection will most likely be in the nanogram range. In some cases, pictograms of target can be detected.

What is the elution volume when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Within practical limits, the elution volume can be scaled up or down to suit your experiment. However, volumes less than 10 µL become more difficult to work with. In addition, the amount of target is important. If you have a lot of beads with a lot of bound target in a small elution volume, your elution may not be very efficient. Typically, 15-100 µL of beads may be eluted in 30 µL. For efficient recovery of the antigen and/or binding partners, the elution volume should at minimum equal the volume of the beads.

How can I quantify the amount of antibody bound to Dynabeads magnetic beads?

There are several methods to quantify the amount of antibody bound to the beads. The crudest method is to measure the concentration of antibody in the coupling reaction before and after antibody attachment. Either fluorescence measurements or absorbance at 280 nm can be used. Alternatively, you could measure the amount of antibody bound to the beads by fluorescence, chemiluminescence, or radiolabeling detection methods. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How long should I incubate my antibody with the lysates?

Incubation time will depend on the immunogenicity of the primary antibody and its binding affinity with the specific antigens. For a good primary antibody, 30-40 minutes incubation should work well. If you are working with a poor antibody or a very low-abundance protein, you could try to increase binding by incubating overnight. However, this also increases the chance of background protein binding.

When should I covalently bind the antibody to the Dynabeads surface?

If the target protein has the same molecular weight as the heavy or light chain antibody, we would recommend covalently binding the antibody to the bead surface. This can be done by either crosslinking the antibody to the Dynabeads Protein A or G magnetic beads, or secondary coated beads, or by using one of the surface-activated Dynabeads magnetic beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What are the general advantages of using Dynabeads magnetic beads for protein isolation?

Using Dynabeads magnetic beads for protein isolation provides several advantages:

-Rapid binding kinetics: since the number of beads per volume for Dynabeads is approximately 1,000 times higher than for the same volume of a Sepharose slurry, the probability for Dynabeads magnetic beads to hit the target is far greater.

-Incubation time: due to the rapid binding kinetics, the protocol is usually very short.
-Low background: due to the rapid binding kinetics and the short incubation time, the background is also very low.
-Trapping of impurities: the beads offer no internal volume for binding or trapping of impurities.
-Low antibody consumption: this is because Dynabeads magnetic beads are nonporous, uniform superparamagnetic, monodispersed, highly crosslinked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The beads are coated with a thin layer of a highly crosslinked polystyrene shell that encases the magnetic material and prevents any leakage from the beads or trapping of ligands in the bead interior. The outer layer also provides a defined surface area for the adsorption or coupling of various molecules such as antibodies. Uniformity of bead size and shape provide consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.
-Reproducibility: due to easier practical handling, such as pipetting. No centrifugation steps or preclearing are required.

Are Dynabeads magnetic beads compatible with dithionite, DTT, and EDTA?

No. Not only is dithionite a reducing agent, but the strong affinity of the dithionite ion for bivalent and trivalent metal cations (M2+, M3+) allows it to enhance the solubility of iron, making it a chelating agent. As a result, the iron in the Dynabeads magnetic beads is reduced and pulled out when they are exposed to dithionite. The same is observed if Dynabeads magnetic beads are exposed to DTT and EDTA. With EDTA, we highly recommend checking the minimal amount of EDTA that your specific molecules would tolerate for binding to the Dynabeads, and if it will affect your specific application. For some applications, low concentrations of EDTA can be tolerated by Dynabeads. On the other hand, using 10 mM EDTA with heating affects the binding of biotin molecules to Dynabeads streptavidin. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Are Dynabeads magnetic beads compatible with Urea?

Yes, they are compatible with 6-8 M Urea when used during post-coupling steps.

Are Dynabeads magnetic beads compatible with centrifugation?

Dynabeads magnetic beads, being magnetic in nature are really not designed to be centrifuged. That being said, the beads themselves are compact, as the pores in the polymer matrix are filled with magnetic material and coated with a final outer polymer shell that will further add to the rigidity of the beads. Hence, pressure should theoretically not be a problem for the beads themselves, but the force exerted by the beads on surrounding cells in the pellet may be detrimental to the cells. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What are the benefits of using magnetic beads in immunoprecipitation (IP) experiments?

Magnetic beads, unlike agarose beads, are solid and spherical, and antibody binding is limited to the surface of each bead. While magnetic beads do not have the advantage of a porous center to increase the binding capacity, they are significantly smaller than agarose beads (1 to 4 µm), which collectively gives them adequate surface area-to-volume ratios for optimum antibody binding.

High-power magnets are used to localize magnetic beads to the side of the incubation tube and out of the way to enable cell lysate aspiration without the risk of also aspirating immune complexes bound to the beads. Magnetic separation avoids centrifugation, which can break weak antibody-antigen binding and cause loss of target protein.

However, an issue with the use of magnetic beads is that bead size variations may prevent all beads from localizing to the magnet. Additionally, while immunoprecipitation using agarose beads only requires standard laboratory equipment, the use of magnetic beads for immunoprecipitation applications requires high-power magnetic equipment that can be cost-prohibitive. Read more about our Magnetic Immunoprecipitation Products (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/immunoprecipitation.html#products).

Which Dynabeads magnetic beads should I use if I am interested in isolating cell organelles with an indirect method?

If you are not concerned about bead release you can use:
- Secondary-coated Dynabeads magnetic beads
- Dynabeads Protein A/G magnetic beads
- Dynabeads Streptavidin magnetic beads

If you would like to remove beads gently after isolation you can use:
- FlowComp Flexi magnetic beads
- Dynabeads CELLection Biotin Binder
- Dynabeads CELLection Pan mouse IgG

Citations for isolation of endosomes and lysosomes:
- Mol Biol Cell 11:2775 (2000)
- J Biol Chem 275:7004 (2000)
- J Cell Sci 111:141 (1998)
- Proc Natl Acad Sci U S A 95:8613 (1998)

Citations reporting the isolation of transport vesicles originating from the ER:
- Science 289:444 (2000)
- Science 295:848 (2002)
- Mol Biol Cell 9:623 (1998)
- J Cell Sci 110:2117 (1997)
- J Neurosci 21:7506 (2001)
- J Biol Chem 274:35577 (1999)

Citations reporting the use of Dynabeads magnetic beads to isolate mitochondria and other organelles:
- Electrophoresis 19:1205 (1998)
- Biochem Biophys Res Commun 295:1027 (2002)
- J Neurosci 21:7506 (2001)
- J Biol Chem 276:32338 (2001)
- J Cell Sci 110:2117 (1997) Jones SM, et al (1998) In: Cell Biology: A laboratory Handbook. Academic Press. pp 12-25
- J Cell Biol 138:291 (1997)
- J Biol Chem 275:7004 (2000)

Citations reporting the use of Dynabeads magnetic beads to isolate mitochondria and chloroplasts and also mitochondrial/chloroplast nucleic acid:
- Genomics 60:67 (1999)
- Biotechniques 26:336 (1999)
- Int J Legal Med 108:237 (1996)
- Am J Hum Genet 60:408 (1997)
- Int J Legal Med 106:85 (1993)
- Biotechniques 16:484 (1994)
- Mol Ecol 4:731 (1995)

What choice of Dynabeads magnetic beads do you offer for isolating organelles if I would like to covalently link my own antibody to the beads?

We have several surface-activated Dynabeads magnetic beads that allow you to couple your own antibody covalently to the beads. These products include M-450 Tosylactivated beads (Cat. No. 14013), M-280 Tosylactivated beads (Cat. Nos. 14203 and 14204), M-450 Epoxy beads (Cat. No. 14011), M-270 Epoxy beads (Cat. No. 14301), M-270 amine beads (Cat. No. 14308), and M-270 carboxylic acid (Cat. No. 14306D).

How do I remove the Dynabeads magnetic beads after isolation of cell organelles?

Dynabeads magnetic beads used for organelle isolation are not designed to elute off the organelles after isolation. When isolating proteins, we recommend using high salt or low pH (boiling in SDS buffer will cause denaturation of the proteins). If you want to try to elute the organelles off, you need to consider the interaction between the organelle and ligand/antibody. If this interaction is strong, you will need more harsh conditions than if the interaction is weaker. High salt may cause the organelles to burst (osmotic stress). You may try low pH, but we can't guarantee the results, and you will need to optimize. We have one product-Dynabeads CELLection Pan Mouse IgG (Cat. No. 11531D), in which a DNA linker connects the antibody (anti-mouse IgG) to the Dynabeads magnetic beads, and this linker can be broken after isolation using a DNase, to free the antibody with isolated organelle. Our FlowComp Flexi Kit (Cat. No. 11061D) can also be used.

How do I determine the bead-to-organelle ratio when isolating cell organelles using Dynabeads magnetic beads?

This will vary a lot depending on cell type, organelle type and abundance, primary antibody and bead type, it is difficult to give very specific recommendations. Most likely you will need to do a titration of bead-to-organelle ratio to optimize the isolation. Use approximately 2 x 10E7 beads per 100-200 µg of unpurified cellular fraction (lysate) diluted in 300 µL buffer (e.g., PBS w/ 2 mM EDTA and 5% FCS or BSA) as a starting point. If your antibody has high affinity, you should be able to achieve a near-complete selection of your target vesicles. Published articles report from 10-20 (e.g., peroxisomes) to hundreds of organelles bound per bead, but this will depend on your antibody and the coating efficiency of the beads.

Which Dynabeads magnetic bead size should I choose for isolation of cell organelles?

If the sample is very viscous, a large bead is preferred (4.5 µm), which has a higher magnetic content and moves more easily to the magnet in very viscous samples. If high binding capacity is preferred, choose the 2.8 µm bead.

Can vesicles/organelles be covalently bound directly onto Dynabeads magnetic beads (without antibody/ligand linker)?

Yes. We recommend using Dynabeads M-270 Epoxy or Dynabeads M-450 Epoxy for this purpose. The reaction chemistry of these particles allows the vesicles/organelles to bind at low temperature (>4 degrees C).

The Dynabeads M-280 Tosylactivated or Dynabeads M-450 Tosylactivated will require a higher reaction temperature, and it is usually desirable to keep the vesicles/organelles at low temperature.

The purified vesicles/organelles bind covalently to the Dynabeads magnetic beads via vesicle/organelle-proteins. We recommend adding a blocking protein (e.g., BSA) after 1 hour of incubation.

How should I prepare the starting cellular material for organelle isolation using Dynabeads magnetic beads?

It is very important to properly homogenize the cells before use. Homogenization may be easier for some samples such as liver, than for other samples such as yeast. If nuclei are broken during homogenization, DNA will be released and may cause clumping of organelles. Organelle clumps in the unpurified cellular material will make the immuno-isolation technique difficult.

Are there any guidelines for picking the right antigen-antibody interaction for organelle isolation using Dynabeads magnetic beads?

It is essential to identify an appropriate antigen and to have a high-affinity antibody that recognizes the antigen in its native, un-denaturated state. The epitope on the subcellular fraction must be free to bind the specific antibody. If the epitope is “buried” within the fraction and not exposed on the surface, it may not be possible to isolate the target organelles.

How do I determine if the direct technique or indirect technique should be used for organelle isolation using Dynabeads magnetic beads?

Due to the speed and efficiency of isolation, the direct technique is recommended. However if the antigen on the target organelles is inaccessible (e.g., buried within the vesicle), the indirect technique may be advantageous.

Can you give me an overview for isolating organelles with Dynabeads magnetic beads?

Several antibodies that recognize antigenic sites on organelles are commercially available. You can choose between two techniques:

- Direct technique: Coat the Dynabeads magnetic beads with your antibody/ligand by incubating the two together for a short incubation period. Then mix your coated beads with your crude fraction. Target organelles bound to the Dynabeads magnetic beads are separated by the use of a magnet, and washing is performed in an equally fast and easy manner.

- Indirect technique: Mix the antibody/ligand with the target-containing crude fraction and incubate to allow them bind. Then add the Dynabeads magnetic beads to the sample, incubate for a short period to allow the target-antibody/ligand complex bind. Separate and wash using a magnet. The indirect technique cannot be used if the target-specific antibody/ligand is to be coated directly onto one of the surface-activated beads.

What are the advantages of using Dynabeads magnetic beads to isolate cell organelles?

In general, all membrane-bound organelles can be isolated using the Dynabeads magnetic beads. In combination with your own organelle-specific antibody, the immuno-magnetic isolation approach has several advantages:

- The method is rapid and gentle.
- Does not require any labor-intensive density gradient and ultracentrifugation
- Organelle populations with different density can be isolated
- Highly pure organelles are obtained


WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.