Learn More
Dynalene Heat Transfer Fluids Dynalene EG, 5 gallon
SDP
Catalog No. NC0981290
Encompass
Dynalene EG is an inhibited technical grade ethylene glycol for use in most industrial applications. It is blended with Dynalene PE-1 inhibitor for pH stability and superior corrosion protection for steel, copper, brass and other metals.
Catalog No. NC0981290 Supplier Dynalene Heat Transfer Fluids Supplier No. DYNALENE EG
Dynalene EG is an inhibited technical grade ethylene glycol for use in most industrial applications. It is blended with Dynalene PE-1 inhibitor for pH stability and superior corrosion protection for steel, copper, brass and other metals.